missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Endotoxin-Free Water
Pierce Endotoxin-Free Water is free of endotoxins and enzymes, including DNAse, RNAse, and proteases. Recommended for the dilution of endotoxin standards and test samples for all endotoxin detection assays and molecular biology or general biochemistry applications. Contains <0.005 EU/mL endotoxin.
Brand: Thermo Scientific™ A43883
10.86 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Pierce Endotoxin-Free Water is free of endotoxins and enzymes, including proteases.Using Endotoxin-Free Water helps eliminate the risk of harmful activity produced by bacteria that can interfere with cell health and function. It is suitable for usage in molecular biology or general biochemistry applications and contains <0.005 EU/mL endotoxin.
Specifications
Endotoxin-Free Water | |
Bottle(s) | |
4 x 50 mL |
<0.005 EU/mL | |
Sterile | |
Sterile filtered with 0.5 μm filter |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction