missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Fab Micro Preparation Kit

Digest human or mouse IgG antibodies with immobilized papain protease and purify Fab fragments using Protein A agarose.

Brand:  Thermo Scientific™ 44685


Product Code. 10321174

  • £481.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Fab Preparation Kits use immobilized papain protease to digest human or mouse IgG antibodies to make separate Fab and Fc fragments and subsequently to purify the Fab using Protein A agarose.

These kits use papain, a nonspecific thiol-endopeptidase, to enzymatically cleave whole IgG just above the hinge region to create two separate Fab fragments and one Fc fragment per antibody molecule. Because the papain protease is supplied in immobilized form as beaded agarose resin, the digestion reaction is easily stopped by removing the resin from the IgG solution; the result is digest products that are enzyme-free. The antibody digestion reaction is performed in convenient disposable spin columns that allow efficient removal of the immobilized protease and maximum recovery of the IgG fragments. Also included in the kits are Protein A spin columns and buffers to efficiently purify the resulting fragments. Protein A binds the Fc fragments and undigested IgG, allowing the pure Fab fragments to be recovered in the flow-through fraction. Thermo Scientific Zeba™ Desalting Spin Columns make IgG-sample preparation quick and easy.

Highlights:

  • Enzyme-free digestion products – immobilized papain (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the protease

  • Suitable for human and other species of IgG – the kit procedure is optimized for human, mouse and rabbit IgG, but papain-based digestion is effective for many species and subclasses of IgG including rat, goat and pig (Note: for best results with mouse IgG1, use Part No. 44980)

  • High capacity – kit provides for digestion and purification from as much as 4mg of IgG at a time. Use the standard size kit for fragmenting 0.25 to 4mg IgG, and the micro kit for 25 to 250μg IgG

  • Provides ready-to-use Fab – digestion and final recovery of purified Fab fragments occurs in neutral pH sodium phosphate buffer, suitable for storage or immediate use in typical applications

  • Complete and easy to use – kits include all reagents, spin columns and accessories needed to prepare and purify antibody fragments

Includes:

Papain agarose resin, cysteine, digestion buffer, Protein A columns with binding and elution buffers, spin desalting columns and accessories

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Pierce Fab Micro Preparation Kit
10 Antibody Samples, each containing 25 to 250 μg IgG
Pierce™
Sufficient For: 10 antibody samples, each containing 25 to 250 μg IgG
• Pierce Papain Agarose, 0.5 mL
• Cysteine-HCl, 1 g
• Fab Digestion Buffer, 55 mL
• NAb Protein A Plus Spin Columns, 0.2 mL, 2 columns
• PBS Packs (each makes 500 mL), 2 packs
• IgG Elution Buffer, 50 mL
• Zeba Desalt Spin Columns, 7K MWCO, 0.5 mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge Tubes, 2 mL, 30 tubes

Store at 4–8°C.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Fab Micro Preparation Kit

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Respiratory sensitiser Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium chloride, Sodium phosphate dibasic, Papain, L-(+)-Cysteine hydrochloridemonohydrate

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.