missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Far-Western Blot Kit for Biotinylated Proteins

Detect proteins in gels or on membranes via interaction with biotin-labeled bait proteins as primary probes.

Brand:  Thermo Scientific™ 23500

Product Code. 10678644

  • £540.00 / 1 set
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Western Blot Kit
Pierce™
Streptavidin-HRP
10 Mini Gels
Chemiluminescence
220 mL
All species
Sufficient For: 10 mini gel or blot experiments with a biotinylated-protein probe
• Streptavidin-HRP Conjugate, 0.1 mg
• Dilution Buffer (10X), 50 mL
• PBS Packs (each makes 500 mL), 17 packs
• Luminol Enhancer, 55 mL
• Stable Peroxide Buffer, 55 mL
• Surfact-Amps 20 Detergent Solution, 6 x 10 mL
• Pre-Cut Cellophane, 10 sheets

Store kit at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZB390271 Certificate TypeCertificate of Analysis Date01/03/2024 Catalog Number23500
Lot NumberYK387279 Certificate TypeCertificate of Analysis Date07/12/2023 Catalog Number23500
Lot NumberYI367052 Certificate TypeCertificate of Analysis Date23/09/2023 Catalog Number23500
Lot NumberXH362405 Certificate TypeCertificate of Analysis Date31/08/2022 Catalog Number23500
Lot NumberWK341704 Certificate TypeCertificate of Analysis Date11/11/2021 Catalog Number23500
Special Offers

Special Offers

  • Multiple Manufacturers
    Save on thousands of eligible products.
    Promo Code: 24111

    Expires2025-03-14

    Days

    4

    Hours

    6

    Minutes

    8

    Seconds

    23
    New Offer
    Save Now
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Far-Western Blot Kit for Biotinylated Proteins > Biotin far-Western kit; 220mL kit

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.