Learn More
Thermo Scientific™ Pierce™ Fast Wash Buffer, 10X
Shop All Thermo Scientific ProductsDescription
Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with Thermo Scientific™ Pierce Fast Blocking Buffer, a ready-to-use blocker solution.
Additional Information and Online Ordering
Highlights:
Fast – use both buffers to shorten the typical Western blot development by over 2 hours
Simple – optimized protocol makes Western blot analysis easier than ever
Low background – provides results comparable to classic Western blotting buffers
Specifications
Specifications
| Chemical Name or Material | Wash Buffer |
| Description | Pierce Fast Wash Buffer, 10X |
| For Use With (Application) | Western Blot |
| Physical Form | Liquid |
| Product Line | Pierce |
| Quantity | 1 L |
Safety and Handling
- Pierce Fast Wash Buffer, 10X
- Warning
- Serious eye damage/eye irritation Category 2
- H319-Causes serious eye irritation.
- P264-Wash hands thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : sodium chloride, trometamol, Poly(oxy-1,2-ethanediyl),,1,3,3-tetramethylbutyl)phenyl]-
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.