Learn More
Thermo Scientific™ Pierce™ Fast Blocking Buffer
Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with this ready-to-use blocker solution.
£161.00 - £282.00
Specifications
Product Type | Blocking Buffer |
---|---|
Form | Liquid |
For Use With (Application) | Western Blot |
Concentration | 1X |
Buffer | TBST |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
13474289
|
Thermo Scientific™
37575 |
500 mL |
£282.00
500mL |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13494289
|
Thermo Scientific™
37576 |
100 mL |
£161.00
100mL |
Estimated Shipment: 16-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Optimize Western blotting systems by decreasing both background signal and incubation time compared to traditional blocking agents with Thermo Scientific™ Pierce Fast Blocking Buffer, a ready-to-use blocker solution.
Additional Information and Online Ordering
Pierce Fast Blocking Buffer streamlines Western blot protocols by providing consistent, high-quality blocking for nitrocellulose or PVDF membranes in only 5 minutes. Blocking and probing steps with Fast Blocking Buffer prevents nonspecific binding of detection antibodies during probing steps but allows specific detection to occur. Pierce Fast Wash Buffer provides efficient membrane washing after incubation with a primary and secondary antibody to effectively shorten total washing time to 20 minutes (from the typical 90 minutes). These two buffers are compatible with antibodies and biotin-avidin systems, as well as both nitrocellulose and PVDF membranes. The blocking buffer provides rapid and effective blocking in just five minutes while producing results comparable to traditional blockers for Western blots.Highlights:
Fast – use both buffers to shorten the typical Western blot development by over 2 hours
Simple – optimized protocol makes Western blot analysis easier than ever
Low background – provides results comparable to classic Western blotting buffers
Specifications
Blocking Buffer | |
Western Blot | |
TBST |
Liquid | |
1X | |
Upon receipt store at 4°C. |
For Research Use Only. Not for use in diagnostic procedures.