Learn More
Thermo Scientific™ Formic Acid, LC-MS Grade
A high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.
Brand: Thermo Scientific™ TS-28905
Code : 16
Additional Details : CAS Number : 64-18-6 Weight : 0.05580kg Transport : UN number : 1779 Chem class : 8 Pack group : II
Description
Thermo Scientific™ Formic Acid is high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.
Formic acid is a component found in reverse-phase mobile phases to provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing reagent, and it does not suppress MS ionization of polypeptides when used as a mobile phase component.
Ordering Alerts:
- X in the ordering table indicates that hazardous shipping charges apply.
Storage Requirements:
- Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Specifications
Formic acid | |
Formic Acid | |
Liquid | |
10 x 1 mL | |
Pierce™ | |
Liquid |
98 to 100% | |
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area | |
Solvent (Pure) | |
99% Formic Acid | |
LC/MS |
Safety and Handling
- formic acid
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P311-Call a POISON CENTER/doctor/ .
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : formic acid