missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Formic Acid, LC-MS Grade

A high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.

Brand:  Thermo Scientific™ 85178

175.65 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.



 View more versions of this product

Product Code. 13454279

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Formic Acid is high-purity solvent supplied in glass ampules as a convenient, contamination-free alternative for preparing elution solvents for HPLC separations of protein and peptides.

Formic acid is a component found in reverse-phase mobile phases to provide protons for LC/MS analysis. The presence of a low concentration of formic acid in the mobile phase is also known to improve the peak shapes of the resulting separation. Unlike trifluoroacetic acid (TFA), formic acid is not an ion-pairing reagent, and it does not suppress MS ionization of polypeptides when used as a mobile phase component.

Highlights:

  • Prescored, nitrogen-flushed, amber glass to protect formic acid from light and moisture
  • 99% purity for consistent LC baselines and no interference introduced into LC and mass spectrometry applications
  • Convenient format simplifies preparation of gradient and isocratic mobile phases containing 0.1% (v/v) formic acid in water or acetonitrile
  • Contents of a single vial in a final volume of 1L solvent yields a mobile phase of the most common formic acid concentration

Ordering Alerts:

  • X in the ordering table indicates that hazardous shipping charges apply.

Storage Requirements:

  • Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Formic Acid
Liquid
50 mL
Store in a cool, dry, and well-ventilated area. Protect from direct sunlight.
Solvent (Pure)
Pierce™
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Formic Acid

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 A

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : formic acid

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.