missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Gaussia Luciferase Glow Assay Kit

Obtain bright and stable bioluminescence signal (half-life greater than 1 hour) in the presence of Gaussia luciferase reporters

Brand:  Thermo Scientific™ 16161

443.52 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 12152240

  • £672.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The Thermo Scientific™ Pierce Gaussia Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life greater than 1 hour), especially in the presence of Gaussia-Dura luciferase reporter.

The Thermo Scientific Pierce Gaussia Luciferase Glow Assay Kit provides a sensitive bioluminescence system to determine expression of a gene of interest with a luciferase reporter. The extremely bright and stable bioluminescence signal (half-life greater than 1 hour) is produced in the presence of the Gaussia-Dura luciferase reporter due to oxidation of the luciferase substrate coelenterazine (Figure 1). Gaussia luciferase is a 20 kDa protein from the marine copepod, Gaussia princeps, that has greater protein stability and signal brightness than firefly and native Renilla luciferase. The bioluminescent enzyme is highly secreted into the cell culture media, allowing for live cell monitoring of reporter activity. Light output generated by the luciferase reaction can be correlated to the amount of Gaussia luciferase protein produced and used to determine the activity of the promoter driving Gaussia expression.

Features of the Gaussia Luciferase Glow Assay Kit:
Sensitive—highly sensitive detection of Gaussia luciferase activity
Stable—increased signal stability
Simple—does not require a luminometer with injectors
Compatible—assay reagents compatible with other Gaussia luciferases
Automation-friendly—amenable to high throughput assays
Convenient—contains a universal cell lysis buffer and optimized glow assay reagent
Safe—allows one to perform non-radioactive assays

When used with Thermo Scientific Gaussia-Dura Luc Vectors, the kit provides an extremely sensitive bioluminescent reporter assay system for detection of secreted or intracellular luciferase activity from promoter or pathway activation with extended signal stability. The light output generated by the luciferase reaction (emission maximum of 485 nm) indicates that Gaussia luciferase was transcribed. This light emission can be correlated to the amount of luciferase protein produced, which in turn is proportional to the activity of the promoter for Gaussia luciferase expression and indicates that the regulatory region of the target gene was active. This reaction does not require adenosine triphosphate (ATP) or other cofactors. The kit includes cell lysis buffer, reaction buffer and substrate and can be used to detect the activity of any Gaussia luciferases or derivatives that use coelenterazine as a substrate.

The Gaussia Luciferase Glow Assay Kit requires:
Gaussia luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Varioskan LUX Microplate Readers.

Applications of the Gaussia Luciferase Glow Assay Kit:
• Promoter studies for analyzing cis- regulatory elements and trans-acting factors
• Drug screening
• siRNA and miRNA screening
• Multiplexed assays to study off-target effects
• Secretory pathway/protein localization reporter assays
• Signal transduction pathway analysis
• RNA splicing studies

The Gaussia-Dura gene used in the Thermo Scientific Gaussia-Dura Luc Vectors is derived from the wild type Gaussia gene and provides equally bright but more stable light output. The Gaussia Glow Assay Kit reagent yields a Gaussia-Dura luciferase reaction with glow-type kinetics that provides a lower intensity but longer-duration signal allowing for higher sample throughput. This glow-type assay with a prolonged luminescence signal does not require reagent injectors and is recommended for luminometers without injectors or for applications requiring batch processing of samples.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Assay Kit
1000 Luciferase Assays
Sufficient For: 1000 luciferase assays in microplate wells
• Gaussia Glow Assay Buffer, 50 mL (store at 4°C)
• 100X Coelenterazine, 0.5 mL (store at -80°C)
• Luciferase Cell Lysis Buffer (2X), 60 mL (store at room temperature)

Kit may be stored at -80°C, or individually at indicated temperatures.
Mammalian Cells
Luminometer (Microplate)
Enzyme Labeled
Enhanced Chemiluminescence
Pierce™
EF-1α
Luciferase, Gaussia Luciferase
1000 Reactions
Reporter Enzyme, Luciferase Reporter Assay
Bioluminescence
384-well plate, 96-well plate
Chemical Substrate
Luciferase Substrate
Coelenterazine
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Gaussia Luciferase Glow Assay Kit

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity Category 2

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H302-Harmful if swallowed.
  • H311-Toxic in contact with skin.
  • H319-Causes serious eye irritation.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
  • H371-May cause damage to organs.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P330-Rinse mouth.
  • P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.