missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ β-Glucan Blocker
Pierce β-Glucan-Blocker is added to the samples to ascertain the validity of the positive control results and rule out false positive results that can come from sample contamination with glucans.
Brand: Thermo Scientific™ A43887
27.76 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.
This item is not returnable.
View return policy
Description
Pierce β-Glucan-Blocker is added to samples suspected of β-1,3-glucans contamination, preventing the occurrence of false positive results in endotoxin detection (LAL) assays. β-1,3-glucans contamination may arise from the presence of yeast and cellulosic materials. β-1,3-glucans, in sufficient amounts, can trigger a LAL response not emanating from an endotoxin-mediated response. This blocker contains <0.005 EU/mL endotoxin.Specifications
Store at 2 to 30°C. |
|
Sterile | |
Vial(s) | |
5 x 10 mL |
Beta-Glucans Blocker | |
<0.005 EU/mL | |
LAL Endotoxin Assay |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction