missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Guardian™ Peroxidase Conjugate Stabilizer/Diluent

Preserve diluted (1-1,000ng/mL) HRP conjugates for more than six months at room temperature without significant loss of activity using Guardian Peroxidase Conjugate Stabilizer/Diluent.

Brand:  Thermo Scientific™ 37548

 View more versions of this product

Product Code. 11849320

  • £149.00 / 200mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Guardian Peroxidase Conjugate Stabilizer/Diluent preserves diluted (1-1,000ng/mL) HRP conjugates for more than six months at room temperature without significant loss of activity.

  • Assay-compatible - simply add your favorite blocking buffer to create the ideal diluent for your HRP-based ELISA system

  • Convenient - store ready-to-use dilutions (1:1,000-1:100,000) that maintain enzyme activity in the refrigerator without aliquoting

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Peroxidase Conjugate Stabilizer/Diluent
Diluent, Stabilizing Reagent
200 mL
Guardian™
Upon receipt store at room temperature or 4°C.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.