missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Heptafluorobutyric Acid (HFBA), Sequencing Grade
Bottles and ampules of 99.5%-pure HFBA (MW 214.04, CAS 375-22-4), valid for use as an ion-pairing agent in reverse-phase peptide separations (HPLC).
Brand: Thermo Scientific™ TS-25003
This item is not returnable.
View return policy
Description
Thermo Scientific™ Pierce Heptafluorobutyric Acid (HFBA) is manufactured and tested to meet strict specifications that ensure superior performance for use as an ion-pairing agent in reverse-phase peptide separations.
Highlights:
- Typical purity is 99.7% by GC; <0.1% water
- Sequencing reagent for classical and automated Edman degradation of peptides and proteins
- Density: 1.645
- B.P. 120°C
- Packaged under nitrogen in amber glass ampules or bottles
- Clear, colorless liquid
Ordering Information:
X in the ordering table indicates that hazardous shipping charges apply.

Specifications
Heptafluorobutyric Acid (HFBA) | |
Liquid | |
100 mL | |
Pierce™ |
Store at room temperature. | |
Ion Pairing Reagent | |
99.5% HFBA |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction