missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A Plus UltraLink™ Resin

Extremely durable polyacrylamide support with densely immobilized Protein A for high-capacity IgG purification.

Brand:  Thermo Scientific™ 53142

364.78 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 11889400

  • £598.00 / 5mL
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Protein A Plus UltraLink Resin is an extremely durable beaded polyacrylamide support to which IgG-binding Protein A has been immobilized at high density for use in IgG purification methods.

The Protein A Plus UltraLink Resin is used for high-capacity binding and purification of IgG in gravity-flow, spin-column or medium-pressure chromatography methods. Protein A is a popular antibody-binding protein that is especially useful for purification of IgG from rabbit or human serum.

UltraLink Resin is an azlactone-activated polyacrylamide support that is hydrophilic, charge-free, high capacity, highly crosslinked, rigid and porous. This support is especially useful for medium pressure techniques when using large sample volumes requiring fast-flow techniques (FPLC) and large-scale applications. Unlike typical beaded agarose supports, UltraLink Support does not crush or collapse when subjected to high flow rates that result in pressures exceeding 25psi.

Highlights:

  • Immobilized Protein A - specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG

  • High capacity - “Plus” variety has a dense load of immobilized Protein A, providing a binding capacity of at least 30mg human IgG per milliliter of resin

  • UltraLink Resin - acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications

Properties of Thermo Scientific UltraLink Resin:

  • Support pH Stability: 1 to 13
  • Average Particle Size: 50 to 80 microns
  • Exclusion Limit: greater than 2,000,000 daltons
  • Average Surface Area: greater than 250m2 per gram of beads
  • Average Pore Volume: greater than 1.2mL per gram of beads (> 60% of bead volume)
  • Pore Size: 500 to 1000 angstroms (50 to 100nm)
  • Maximum Linear Velocity: 3000cm per hour
  • Maximum Pressure: 100psi (6.9 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Resin
UltraLink Affinity Resin
Protein A
Upon receipt store products at 4°C. Products are shipped at ambient temperature.
Pierce™, UltraLink™
Bottle
IgG Antibodies
5 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.