missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein G Plus Agarose

High-capacity, recombinant Protein G beaded agarose resin for a variety of antibody affinity purification methods.

Brand:  Thermo Scientific™ 22852

856.26 GBP valid until 2024-12-20
Use promo code "21651" to get your promotional price.


 View more versions of this product

Product Code. 11871325

  • £1206.00 / 10mL
Estimated Shipment: 29-11-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Protein G Plus Agarose is a high-capacity Protein G beaded agarose resin for use in a variety of antibody affinity purification methods.

Pierce Protein G Plus Agarose consists of recombinant Protein G that has been covalently immobilized at high density onto high-quality crosslinked 6% beaded agarose (CL-6B). The resin is effective for affinity purification of IgG from serum and other fluids of many mammalian species. Protein G is especially well suited for use with mouse antibodies (including mouse IgG1) in addition to most IgG isotypes from human, goat and sheep samples.

Highlights:

  • Protein G – immobilized Protein G is ideal for polyclonal IgG purification from mouse, human, cow, goat and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Inert and stable – superior manufacturing method immobilizes Protein G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • High capacity – “Plus” variety of Pierce Protein G Agarose has a dense load of immobilized Protein G, providing a binding capacity greater than 20mg human IgG/mL resin

Properties of crosslinked 6% beaded agarose (CL-6B):

  • Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
  • Average Particle Size: 45 to 165 microns
  • Exclusion Limit: 10,000 to 4,000,000 daltons
  • Maximum Volumetric Flow Rate: approx. 1mL/minute (for 1cm diameter column)
  • Maximum Linear Velocity: 30cm per hour
  • Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Agarose
Agarose Resin, Affinity
Protein G
Upon receipt store product at 4°C. Product is shipped at ambient temperature.
Pierce™
Bottle
IgG Antibodies
10 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.