missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein G Plus UltraLink™ Resin

Extremely durable polyacrylamide support with densely immobilized, recombinant Protein G for various IgG purification methods.

Brand:  Thermo Scientific™ 53128

Product Code. 11839400

  • £560.00 / 2mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Resin
UltraLink Affinity Resin
Protein G
Upon receipt store product at 4°C. Products are shipped at ambient temperature.
Pierce™, UltraLink™
Bottle
IgG Antibodies
2 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZB371260 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number53128
Lot NumberXG357239 Certificate TypeCertificate of Analysis Date16/07/2022 Catalog Number53128
Lot NumberWJ335606 Certificate TypeCertificate of Analysis Date09/11/2021 Catalog Number53128
Lot NumberWC313040 Certificate TypeCertificate of Analysis Date31/03/2021 Catalog Number53128
Lot NumberUI285121 Certificate TypeCertificate of Analysis Date21/09/2019 Catalog Number53128
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Protein G Plus UltraLink™ Resin > Resin; 2mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.