Learn More
Thermo Scientific™ Pierce™ Protein L Plus Agarose
Shop All Thermo Scientific ProductsDescription
Perform a variety of antibody affinity purification methods with Thermo Scientific™ Pierce High-Capacity Protein L Agarose, a high-capacity Protein L beaded agarose resin.
Highlights:
Protein L – immobilized Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
Inert and stable – superior manufacturing method immobilizes Protein L by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
High capacity – “Plus” variety of Pierce Protein L Agarose has a dense load of immobilized Protein L, providing a binding capacity of 10 to 20mg human IgG/mL resin
Properties of crosslinked 6% beaded agarose (CL-6B):
- Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
- Average Particle Size: 45 to 165 microns
- Exclusion Limit: 10,000 to 4,000,000 daltons
- Maximum Linear Velocity: 30cm per hour
- Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather
- than the pressure drop across the column.)
Specifications
Specifications
| Product Line | Pierce |
| Column Type | Agarose Resin, Affinity |
| Format | Bottle |
| Stationary Phase | Protein L |
| Purification Target | Ig Antibodies |
| Content And Storage | Upon receipt store product at 4°C to 8°C. Product is shipped at ambient temperature. |
| Quantity | 2 mL |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.