Learn More
Thermo Scientific™ Hydrochloric Acid Solution, Sequencing Grade
Constant-boiling 6N HCl solution, supplied in 1mL ampules, used to acid-hydrolyze proteins (protein hydrolysis) for HPLC amino acid composition analysis.
Brand: Thermo Scientific™ 24308
Code : A0
Additional Details : CAS Number : 7647-01-0 Weight : 0.01000kg Transport : UN number : 1789 Chem class : 8 Pack group : III
Description
Thermo Scientific™ Pierce Hydrochloric Acid Solution is a constant-boiling 6N HCl solution, supplied in 1mL ampules, that is specifically formulated as a sequencing-grade reagent for total protein hydrolysis for amino acid analysis.
Highlights:
- Hydrolyzes peptides in 6 hours at 150°C
- Specially purified to give ninhydrin-negative blank on hydrolysis
- Packaged in prescored ampules to eliminate contamination and ensure product integrity
Ordering Information:
X in the ordering table indicates that hazardous shipping charges apply.
Specifications
Hydrochloric Acid Solution | |
No product PDF available for storage information. | |
10 x 1 mL | |
Solid |
Safety and Handling
- Hydrochloric Acid Solution, Sequencing Grade
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Hydrogen chloride
For Research Use Only. Not for use in diagnostic procedures.