missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ IP Lysis Buffer

Product Code. p-4531876 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
This item is not returnable. View return policy

Product Code. 11820084

Brand: Thermo Scientific™ 87787

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximize cell lysate yield, purity and compatibility with this immunoprecipitation-optimized protein extraction reagent.

Thermo Scientific™ Pierce IP Lysis Buffer is optimized for cell lysate yield, purity and compatibility with immunoprecipitation.

The IP Lysis Buffer is a mammalian whole cell lysis reagent based on a modified RIPA buffer formulation without SDS. This moderate-strength lysis buffer effectively solubilizes cellular proteins but does not liberate genomic DNA or disrupt protein complexes like ordinary RIPA buffer. Pierce IP Lysis Buffer is specially formulated for pulldown and immunoprecipitation assays.

Highlights:

  • Optimized for compatibility with immunoprecipitation and pull-down assays
  • Compatible with protein assays, reporter assays and immunoassay procedures
  • Ready-made formula is effective for extracting cytoplasmic, membrane and nuclear proteins
  • Gentle formulation helps maintain protein complexes for co-immunoprecipitation (co-IP)
  • Does not liberate genomic DNA which can cause high sample viscosity
TRUSTED_SUSTAINABILITY

Specifications

Quantity 100 mL
Format Liquid
Product Type IP Lysis Buffer
Volume (Metric) 100 mL
Content And Storage Upon receipt store at 4°C.
Product Line Pierce

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.