Learn More
Thermo Scientific™ KMUH (N-κ-maleimidoundecanoic acid hydrazide)
Crosslink sulfhydryls to periodate-treated glycoprotein sugars with this long (19.0A) crosslinker containing maleimide and hydrazide groups.
Brand: Thermo Scientific™ 22111
Code : PN
Additional Details : Weight : 0.80000kg
Description
Thermo Scientific™ Pierce KMUH is a long, maleimide-and-hydrazide crosslinker for conjugating sulfhydryls (cysteines) to carbonyls (aldehyde or ketones, such as those formed by oxidation of glycoprotein carbohydrates).
Highlights:
- Long, noncleavable, aliphatic spacer
- Hydrazide group covalently couples to oxidized carbohydrate residues in glycoproteins and other glycoconjugates
- Reactive groups: maleimide and hydrazide
- Reactive toward: sulfhydryl and carbonyl (aldehyde) groups (or carboxyl groups when used with EDC)
Specifications
Carbohydrate-Sulfhydryl | |
KMUH | |
No | |
Upon receipt store desiccated at 4°C. | |
Ambient | |
Chemical Labeling | |
Amine, Hydrazide, Maleimide | |
Solid | |
DMF, DMSO | |
No |
No | |
409.4 | |
19.0 Å | |
Yes | |
Pierce™ | |
Heterobifunctional | |
Medium (10 to 30 Å) | |
50 mg | |
Standard | |
Crosslinker |
For Research Use Only. Not for use in diagnostic procedures.