Learn More
Thermo Scientific™ Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture)
Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven HeavyPeptide AQUA Ultimate peptides provided at five dilutions distinguished by differential isotopic labeling to assess sensitivity and dynamic range of LC-MS/MS systems.
Brand:  Thermo Scientific™ A40010
 View more versions of this product
Product Code. 16261851
  • £201.00 / 25µL
Explore more special offers
Description
Description
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) contains seven PRTC HeavyPeptide AQUA Ultimate peptides in five isotopologue versions for the generation of seven standard curves to assess sensitivity and dynamic range of LC-MS/MS systems.
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mix) contains 7 of 15 peptides from the Pierce Peptide Retention Time Calibration Mixture (Cat. No. 88320). Each of the seven peptides has five versions (amino acids labeled with 0, 1, 2, 3, or 4 heavy isotopes) at five distinct concentrations. The 7 x 5 mix can be used with Skyline software to assess dynamic range, linearity, and lower limit of quantitation (LLOQ).
The Pierce LC-MS/MS System Suitability Standard (7 x 5 mixture) is:
High quality—contains five sets of seven HeavyPeptide AQUA Ultimate peptides, each distinguishable through unique isotopologue labeling
Optimized—enables scheduling of MS acquisition windows for improved quantification and increased multiplexing
Quantitative—assesses dynamic range of instrument and instrument sensitivity
Predictive—enables assessment of chromatography and MS instrument performance
Applications:
• Optimization of LC & MS parameters
• Standard for data-dependent acquisition (DDA)
• Standard for targeted MS (e.g., parallel-reaction monitoring (PRM)
• Identification of total peptide elution window
• Assessment dynamic range of nano or capillary flow LC-MS/MS systems
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture) (0.5 pmol/μL)
25 μL
Mass Spectrometer
Dry Ice
0.5 pmol/μL
LC-MS/MS System Standard
Pierce™
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Pierce™ LC-MS/MS System Suitability Standard (7 x 5 mixture) > 0.5 pmol/μL, 25 μL