Learn More
Thermo Scientific™ Pierce™ LDS Sample Buffer, Non-Reducing (4X)
Nonreducing lithium dodecyl sulfate sample loading buffer, a unique alternative to homemade and other commercial gel-loading dyes.
Brand: Thermo Scientific™ 84788
33.24 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
Description
Thermo Scientific™ Pierce LDS Sample Loading Buffer (4X) is a nonreducing lithium dodecyl sulfate sample loading buffer optimized for use with Pierce Protein SDS-PAGE Gels.
LDS Sample Buffer, Non-Reducing (4X) is a convenient sample buffer for use in SDS-polyacrylamide gel electrophoresis (SDS-PAGE). The buffer contains coomassie dye, enabling visualization of the electrophoretic progress by the location of the dye front. The LDS Sample Buffer, Non-Reducing (4X) may be used in denaturing gels and is compatible with both Coomassie and silver staining and Western blotting applications.Highlights:
Concentrated – 4X formulation provides more versatility than traditional 2X buffers
Nonreducing – ready to use for non-reducing SDS-PAGE, or the preferred type and amount of reducing agent (e.g., DTT) can be added to produce reducing conditions
SDS alternative – lithium dodecyl sulfate (LDS) replaces sodium dodecyl sulfate (SDS) of traditional loading buffers but functions equally well
Physiologic pH – buffered with triethanolamine (pH 7.6) rather than Tris (pH 6.8)
Lane marker dyes – includes trace amounts of phenol red and coomassie G250 as visible markers of the dye front
Convenient – stable at room temperature, enabling storage at the bench where electrophoresis is performed
Specifications
Upon receipt store at room temperature. | |
4X | |
LDS Sample Buffer | |
Pierce™ |
Sample Loading Buffers | |
SDS-PAGE | |
SDS-PAGE Gels | |
5 mL |
Safety and Handling
- LDS SAMPLE BUFFER NON-REDUCING
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
- MIXTURE LIST-Contains : Triethanolamine, Sulfuric acid, monododecyl ester, lithium salt, Hydrogen chloride
For Research Use Only. Not for use in diagnostic procedures.