Learn More
Thermo Scientific™ Pierce™ LTQ ESI Positive Ion Calibration Solution
Calibrate LTQ, LTQ Orbitrap, LXQ and LCQ FLEET mass spectrometers using a mixture of highly purified ionizable molecules designed for positive mode calibration.
Brand: Thermo Scientific™ 88322
247.43 GBP valid until 2024-06-30
Use promo code "21742" to get your promotional price.
Code : PN
Additional Details : Weight : 0.01000kg Transport : UN number : 1992 Chem class : 3 Pack group : II
Description
Thermo Scientific™ Pierce LTQ ESI Positive Ion Calibration Solution is a mixture of highly purified ionizable molecules specifically designed for positive mode calibration of Thermo Scientific™ ion trap and orbitrap instruments.
The Pierce LTQ ESI Positive Ion Calibration Solution is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific Mass Spectrometers. The LTQ ESI Positive Ion Calibration Solution is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications. The stable solution is provided in a leak-proof, high-purity PTFE bottle.Highlights:
Strong peaks – mixture of caffeine, MRFA and Ultramark 1621 in a acetonitrile/methanol/acetic solution
Ready-to-use – load the mass reference standard into a syringe and inject into the instrument
High-purity – mass spectrometry grade reagents in a non-leachable container
Stable – store at room temperature for more than one year
Specifications
Mass Spectrometer | |
Pierce™ | |
Peptides | |
Mass Spectrometry | |
Liquid |
Calibration Solution | |
Store between 2°C and 8°C. | |
Peptides | |
Calibration |
Safety and Handling
- LTQ ESI POSITIVE LON CAL. SOL.
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H319-Causes serious eye irritation.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P233-Keep container tightly closed.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P242-Use only non-sparking tools.
- P243-Take precautionary measures against static discharge.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P271-Use only outdoors or in a well-ventilated area.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P311-Call a POISON CENTER/doctor/ .
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P330-Rinse mouth.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P403+P235-Store in a well-ventilated place. Keep cool.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetic acid, methanol, acetonitrile
For Research Use Only. Not for use in diagnostic procedures.