missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ beta-Galactosidase Assay Reagent

Lyse cultured mammalian cells and measure beta-galactosidase activity using compatible M-PER Cell Lysis Reagent and colorimetric b-Gal Assay Reagent.

£133.00 - £281.00

Specifications

Assay Reporter Enzyme
Content And Storage Store at -20°C.
Detection Method Colorimetric
Form Liquid
For Use With (Application) Absorbance
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10638844
View Documents
Thermo Scientific™
75705
25 mL
£133.00
25mL
Estimated Shipment: 07-01-2025
to see stock.
Add to basket
 
11849420
View Documents
Thermo Scientific™
75710
3 x 25 mL
£281.00
Pack of 3
Estimated Shipment: 03-01-2025
to see stock.
Add to basket
 
Description

Description

Perform a simple colorimetric method for lysing cultured mammalian cells and measuring beta-Gal activity using Thermo Scientific™ Mammalian β-Galactosidase Assay Kit.

The Assay Reagent and Kit comprise a simple and versatile assay system for measuring β-galactosidase activity. Mammalian cells grown on a plate may be lysed and tested for β-galactosidase activity by adding a single, all-in-one lysis-and-assay solution. Alternatively, the included Thermo Scientific M-PER™ Mammalian Protein Extraction Reagent may be used to prepare lysate from washed adherent or pelleted cells, and then a portion of the lysate used to quantitate the β-Gal activity. With M-PER Reagent, there is no need to scrape cells or to perform freeze-thaw cycles. The remaining lysates prepared with M-PER Reagent may be used for applications such as reporter assays (luciferase, β-galactosidase and chloramphenical acetyltransferase), protein kinase assays (PKA, PKC, and Tyrosine Kinase), immunoassays (Western blot, ELISA, RIA) and protein purification.

Highlights:

  • Choose stand-alone assay reagent or complete kit for cell lysis and detection
  • Colorimetric assay easily measured in a plate reader or spectrophotometer (405nm)
  • Adaptable to 96-well microplates or tube assay formats
  • Perform homogeneous screening assays or measure activity secondarily after cell lysis
  • Also compatiible with prepared lysates from other species such as bacteria

Includes:

Stand-alone Thermo Scientific Pierce™ β-Galactosidase Assay Reagent, Thermo Scientific M-PER Mammalian Protein Extraction Reagent (Part No. 78503), and ready-to-use Stop Solution

Specifications

Specifications

Reporter Enzyme
Colorimetric
Absorbance
Beta-Gal Substrate
Beta-Galactosidase
Microplate Reader
Pierce beta-Galactosidase Assay Reagent
Store at -20°C.
Liquid
Pierce™
Chemical Substrate
96-well Plate
Enzyme Labeled
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.