missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Mass Spec Sample Prep Kit for Cultured Cells

Product Code. 13454219 Shop All Thermo Scientific Products
Click to view available options
Quantity:
20 Reaction Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 13454219

Brand: Thermo Scientific™ 84840

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Perform preparation of clean peptide mixtures from cultured cells for mass spectrometry (MS) analysis with this easy-to-use, comprehensive kit.

Perform preparation of clean peptide mixtures from cultured cells for mass spectrometry (MS) analysis with Thermo Scientific™ Pierce Mass Spec Sample Prep Kit for Cultured Cells, an easy-to-use, comprehensive kit.

Additional Information and Online Ordering

The Pierce Mass Spec Sample Prep Kit for Cultured Cells contains all the necessary reagents and enzymes to prepare up to 20 samples (1 million cells each) for MS analysis. The simple and robust workflow uses a lysis protocol that generates approximately 100μg of protein per sample and includes a control digestion indicator to monitor the efficiency of the two-step enzymatic digestion protocol. The kit procedure, which includes a Digestion Indicator as an internal protein/peptide control, produces consistently and reproducibly clean peptide mixtures for protein identification.

Highlights:

  • Complete – includes all reagents, a digestion indicator control, proteases and an optimized protocol needed to process up to 20 samples

  • Simple – user friendly kit can provide reproducible results even for non-expert MS analysts

  • Flexible – can be adapted to handle sample sizes between 10 to 200μg

  • High yield – total protein yield from 1 million cells is greater than 100μg

  • Optimized – cysteine reduction and alkylation are 100% with less than 1% over alkylation of non-cysteine residues

  • Efficient - percentage of missed cleavages is less than 10%

  • Compatible – final preparation is ready for direct MS analysis and other downstream applications, including mass-tag labeling

Includes:

Kit contains proprietary lysis and digestion buffers, digestion indicator, DTT, iodoacetamide, Lys-C, trypsin

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Equipment) Microcentrifuge, Mass Spectrometer
Product Type Sample Preparation Kit
Product Line Pierce
Content And Storage Upon receipt, remove Insert A (containing Pierce Digestion Indicator, Lys-C Protease and Pierce Trypsin Protease, MS Grade) and store at -20°C. Store the remaining components at 4°C. Product is shipped on dry ice.
Starting Material Mammalian Cells
Final Product Type Peptide
Detection Method Mass Spectrometry
Workflow Step Protein Digestion, Cell Lysis, Sample Clean Up
Quantity 20 Reaction Kit
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.