missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ 16% Formaldehyde (w/v), Methanol-free

Vials of high-quality, methanol-free formaldehyde, a general IHC fixative and reversible amine-crosslinking agent for protein and nucleic acid samples.

Brand:  Thermo Scientific™ 28906

41.65 GBP valid until 2024-12-31
Use promo code "22915" to get your promotional price.



Code : PN

Additional Details : CAS Number : 50-00-0 Weight : 1.02060kg


 View more versions of this product

Product Code. 10751395

  • £46.40 / Pack of 10
In Stock
to see stock.

Description

Description

Thermo Scientific™ Pierce Formaldehyde Ampules are vials of high-quality, methanol-free formaldehyde for use as a general immunohistochemistry fixative and reversible amine-crosslinking agent for protein and nucleic acid tissue samples.

Formaldehyde is a highly reactive, cell-permeable agent that is used as a reversible cross-linking agent for proteins and nucleic acids within the cell or as a general cell-fixing agent for imaging-based applications. Cell permeability allows the reagent to readily enter living cells, enabling intracellular applications, such as protein interaction discovery or bioimaging. Formaldehyde produces reversible cross-links with protein and nucleic acids by coupling neighboring primary amines. Cross-links can be reversed simply by heating the sample.

Highlights:

  • Pure and stable - prepared directly from pure granular paraformaldehyde and sealed in ampules under an inert atmosphere, ensuring that it is methanol-free and resistant to oxidation

  • Versatile - use to cross-link molecules in protein-protein and protein-nucleic acid interaction discovery, bioimaging and flow cytometry

  • Aliquot packaging - choose 1mL or 10mL ampules suitable for low- and high-volume applications ranging from culture-targeted cell fixing to in-cell protein cross-linking

  • Convenient starting concentration - 16% (w/v) formaldehyde solution in ultrapure water simplifies dilution to the required working concentration

Specifications

Specifications

Formaldehyde
Liquid
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area.
Pierce
Fixation Reagent
10 x 1 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 16% Formaldehyde (w/v)Methanol-free, 10 x 1 mL

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H317-May cause an allergic skin reaction.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H351-Suspected of causing cancer.

Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P201-Obtain special instructions before use.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

Supplemental information
  • MIXTURE LIST-Contains : Formaldehyde

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.