Learn More
Thermo Scientific™ Micro BCA™ Reagent A (MA)
Shop All Thermo Scientific ProductsDescription
Thermo Scientific™ Pierce Micro BCA Protein Assay uses a concentrated form of the BCA Reagents to accurately measure dilute protein solutions (0.5 to 20μg/mL).
Additional Information and Online Ordering
Highlights:
Sensitive – accurately detect down to 0.5μg/mL (2μg/mL in microplate format)
High linearity – linear working range for BSA equals 0.5 to 20μg/mL
Colorimetric – measure with a standard spectrophotometer or plate reader (562nm)
Compatible – unaffected by typical concentrations of most ionic and nonionic detergents
Convenient – microplate and cuvette protocols provided with the instructions
Stable – kit stable at room temperature; prepared working reagent stable for to 24 hours
Includes:
Three Micro BCA Reagents (MA, MB, MC) and 10 ampules of Albumin Standard (2mg/mL)
Specifications
Specifications
| Content And Storage | Upon receipt store product at room temperature. Product shipped at ambient temperature. Note: If Reagent MA precipitates upon shipping in cold weather or during long-term storage, dissolve precipitates by gently warming and stirring the solution. Discard any reagent that shows discoloration or evidence of microbial contamination. |
| Description | Pierce Micro BCA Reagent A (MA) |
| Assay | BCA Assay |
| Detection Method | Colorimetric |
| Product Line | Micro BCA, Pierce |
| Product Type | Protein Quantitation Assay |
| Specificity | Not Target-Specific |
| Sufficient For | 3200 Microplate Assays |
| For Use With (Application) | Solution-based Detection, Absorbance |
| For Use With (Equipment) | Spectrophotometer, Microplate Reader |
| Show More |
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.