Learn More
Thermo Scientific™ Pierce™ Mini Gel Power Staining Kit
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
Brand: Thermo Scientific™ 22840
129.28 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 2.00000kg
Description
Get fast Coomassie staining of protein gels with Thermo Scientific™ Pierce™ Power Staining Kits, optimized to work exclusively with the Pierce Power Stain Cassette and Pierce Power Station.
Highlights:
- High performance – performance is equivalent to, or better than, traditional Coomassie staining techniques
- Fast – optimized ionic solutions enable Coomassie staining of protein and gel destaining within 6 to 11 minutes when used with the Pierce Power Stainer
- Compatible – verified to work with commonly used precast and homemade SDS-PAGE gels
- Pierce Power Stain Solution, Destain Solution and Gel Pads
- Coomassie staining and destaining for protein visualization in polyacrylamide gels (requires Pierce Power Stain Cassette and Pierce Power Station)
- Pierce Power Stain Cassette and Pierce Power Station or Pierce G2 Fast Blotter (upgraded) with Power Stain Cassette
Specifications
Note: The kit contains sufficient reagents to stain 30 mini-sized gels at 15 mL each. Contents: Mini Staining Gel Pads, 2 boxes of 30 each (8 sheets/pad) Pierce Power Stain Solution, 480 mL Destain Solution, 480 mL |
|
In-Gel Detection | |
Coomassie | |
Proprietary gel stain and destain solutions | |
1 Kit: 60 Pads, 30 Gels | |
Protein |
Mini Gel Power Staining Kit, 30 Gels, Coomassie Dye, Protein Gel Electrophoresis Application, Colorimetric Detection Method, In-Gel Detection, The Kit Contains Sufficient Reagents To Stain 30 Mini-Sized Gels at 15mL Each | |
Colorimetric | |
Fast Protein Gel Stain | |
Pierce™ | |
30 mini-sized gels at 15mL each |
For Research Use Only. Not for use in diagnostic procedures.