missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Mouse IgG1 Fab and F(ab')2 Preparation Kit

Efficiently digest mouse IgG1 antibodies into Fab or F(ab')2 fragments with immobilized ficin protease.

Brand:  Thermo Scientific™ 44980


Product Code. 10781775

  • £724.00 / 1 set

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce Mouse IgG1; Fab and F(ab')2 Preparation Kits use immobilized ficin protease to efficiently digest mouse IgG1 into Fab or F(ab')2 fragments, depending on the chosen reaction conditions.

The kits use ficin protease to enzymatically digest the Fc portion of whole IgG to yield Fab or F(ab')2 fragments. Because the ficin protease is supplied in immobilized form as beaded agarose resin, the digestion reaction is easily stopped by removing the resin from the IgG solution; the result is digest products that are enzyme-free. An optimized procedure and high-performance desalting and Protein A spin columns make these kits easy to use.

Highlights:

  • Enzyme-free digestion products – immobilized ficin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the protease

  • Optimized for mouse IgG1 – the kit procedure is specifically optimized for monoclonal mouse IgG1 antibodies, providing the best possible yield for this antibody subclass

  • High capacity – kit provides for digestion and purification from as much as 4mg of IgG at a time. Use the standard size kit for fragmenting 0.25 to 4mg IgG, and the micro kit for 25 to 250μg IgG

  • Complete and easy to use – kits include all reagents, spin columns and accessories needed to prepare and purify antibody fragments

Includes:

Ficin agarose resin, cysteine, digestion buffer, Protein A columns with binding and elution buffers, spin desalting columns and accessories

TRUSTED_SUSTAINABILITY
Specifications

Specifications

2.5 mL kit
Pierce™
Sufficient For: 10 antibody samples, each containing 0.25 to 4 mg IgG
• Pierce Ficin Agarose, 2.5 mL
• Cysteine-HCl, 1 g
• Mouse IgG1 Digestion Buffer, 120 mL
• NAb Protein A Plus Spin Column, 1 mL, 1 column
• Protein A Binding Buffer, 120 mL
• IgG Elution Buffer, 120 mL
• Zeba Desalt Spin Columns, 7K MWCO, 2 mL, 10 columns
• Spin Columns, 0.8 mL, 10 columns
• Microcentrifuge tubes, 2 mL, 30 tubes

Store at 4–8°C.
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce Mouse IgG1 Fab and F(ab')2 Preparation Kit

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash hands thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : Sodium chloride, L-(+)-Cysteine hydrochloride monohydrate

SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.