missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ MS-Compatible Magnetic IP Kits

Product Code. p-7225608 Shop All Thermo Scientific Products
Click to view available options
Description:
Protein A/G Magnetic Beads
Streptavidin Magnetic Beads
Unit Size:
40 reactions
This item is not returnable. View return policy

Product Code. 15312627

Brand: Thermo Scientific™ 90409

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Achieve highly efficient, compatible immunoprecipitation and co-immunoprecipitation of target antigens for downstream LC-MS analysis using Thermo Scientific Pierce MS-Compatible Magnetic IP Kits with streptavidin or Protein A/G magnetic beads.

Immunoprecipitation is often necessary for the detection and quantitation of low-abundance proteins or rare signaling events. Mass spectrometry analysis relies on two methods for the detection of low-abundance proteins: single reaction monitoring (SRM) and parallel reaction monitoring (PRM), which enable isolation, scheduling, and quantitation of selected proteotypic peptides. These IP kits have been optimized for efficient immunoprecipitation and co-immunoprecipitation of target proteins, and are directly compatible with downstream MS sample preparation and analysis.

MS-compatible—reagents directly compatible with in-solution peptide digestion
Sensitive—procedure successfully enriches low-abundance proteins (low ng range)
Low background—enriched samples contain minimal detergent residuals
Reduced antibody contamination—antibody biotinylation with streptavidin enrichment reduces contamination in the eluate when compared to other IP methods
Robust—procedure and reagents have been verified with numerous targets to ensure consistent enrichment of low-abundance proteins (ng range) with at least two peptides identified per protein

The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Protein A/G) uses high-quality Pierce Protein A/G Magnetic Beads to provide wider flexibility of antibody capture than either Protein A or G alone. The Thermo Scientific Pierce MS-Compatible Magnetic IP Kit (Streptavidin) uses high-quality Pierce Streptavidin Magnetic Beads for use with biotinylated antibodies that may be purchased commercially or labeled using the Pierce Antibody Biotinylation Kit for IP. After the immunoprecipitation procedure, the target-enriched elution fraction is ready for in-solution tryptic digestion for LC-MS analysis without the need for gel purification, detergent removal, or desalting. Additionally, the reagents and procedures have been validated using both manual and automated magnetic separation.

Applications
• Enrichment of low-abundance targets to improve MS sensitivity
• Identification of target proteins that are not detectable by western blotting
• Quantitation of target proteins by LC-MS analysis
• Screening of antibodies for IP-MS

TRUSTED_SUSTAINABILITY

Specifications

Description Protein A/G Magnetic Beads
For Use With (Equipment) KingFisher™ Flex, Magnetic Stand
Product Type Magnetic IP Kit
Product Line Pierce
Sufficient For 40 Reactions
Content And Storage Protein A/G Magnetic Beads, 1 mL
IP-MS Cell Lysis Buffer, 100 mL
IP-MS Wash Buffer A, 75 mL
IP-MS Wash Buffer B, 40 mL
IP-MS Elution Buffer, 6 mL
MgCl2 (1 M), 0.75 mL

Store in refrigerator (2° to 8°C).
Starting Material Protein Samples, Cell Lysate
Final Product Type Protein
Detection Method Mass Spectrometry
Format Kit
Workflow Step Sample Enrichment
Quantity 40 reactions kit
For Use With (Application) Mass Spectrometry
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.