missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ NHS-Activated Magnetic Beads

Immobilize proteins or other ligands for magnetic separation affinity purification with these magnetic particles coated with amine-reactive NHS-ester groups.

Brand:  Thermo Scientific™ 88827

492.66 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.



 View more versions of this product

Product Code. 12196490

  • £966.00 / 5mL
Estimated Shipment: 02-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ Pierce NHS-Activated Magnetic Beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use in custom affinity purification experiments.

The activated magnetic beads contain N-hydroxysuccinimide (NHS) functional groups that react with primary amines forming stable amide linkages. Once they are covalently attached, the immobilized proteins are highly resistant to leaching from the bead surface. When prepared beads are used in experiments, nonspecific binding is negligible because nonreacted NHS-ester groups are thoroughly blocked during the coupling procedure. Pierce NHS-Activated Magnetic beads can be coupled and processed either manually with a magnetic stand or with automated platforms such as the Thermo Scientific KingFisher Instruments.

Highlights:

  • High capacity at least four times higher binding capacity than NHS-activated magnetic beads from other suppliers

  • Easy to use immobilize in a simple one-step reaction with minimal hands-on time

  • Safe no hazardous chemicals needed (e.g., sodium cyanoborohydride and cyanogen bromide)

  • Ligand-compatible use to immobilize with nearly any primary amine-containing compound or affinity ligand

  • Low non-specific binding the bead surface is pre-blocked and any nonreacted NHS-ester groups are fully quenched

  • Protocol-compatible protein coupling to the beads and downstream applications can be performed manually or by automation (e.g., Thermo Scientific KingFisher Instruments)

Recommended for:

Immobilization of ligands for the purification of recombinant proteins; Immobilization of antibodies for immunoprecipitations and co-immunoprecipitations free of antibody contamination in the eluates; Immobilization of secondary antibody for the affinity purification of antibody subtypes from ascites, serum and cell culture supernatant

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Upon receipt store at 4°C. Product shipped with an ice pack.
Liquid
5 mL
Amine
NHS Ester
Pierce™
Recombinant
Magnetic Affinity Bead
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • Pierce NHS-Activated Magnetic Beads

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B

Hazard Statement
  • H222-Extremely flammable aerosol.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H360FD-May damage fertility. May damage the unborn child.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : N,N-dimethylacetamide

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.