Learn More
Thermo Scientific™ Pierce™ NHS-Activated Magnetic Beads
Immobilize proteins or other ligands for magnetic separation affinity purification with these magnetic particles coated with amine-reactive NHS-ester groups.
Brand: Thermo Scientific™ 88826
150.96 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.
View more versions of this product
Description
Thermo Scientific™ Pierce NHS-Activated Magnetic Beads enable covalent, amine-based conjugation of proteins to magnetic beads in a simple mix-and-go format for use in custom affinity purification experiments.
The activated magnetic beads contain N-hydroxysuccinimide (NHS) functional groups that react with primary amines forming stable amide linkages. Once they are covalently attached, the immobilized proteins are highly resistant to leaching from the bead surface. When prepared beads are used in experiments, nonspecific binding is negligible because nonreacted NHS-ester groups are thoroughly blocked during the coupling procedure. Pierce NHS-Activated Magnetic beads can be coupled and processed either manually with a magnetic stand or with automated platforms such as the Thermo Scientific KingFisher Instruments.Highlights:
High capacity at least four times higher binding capacity than NHS-activated magnetic beads from other suppliers
Easy to use immobilize in a simple one-step reaction with minimal hands-on time
Safe no hazardous chemicals needed (e.g., sodium cyanoborohydride and cyanogen bromide)
Ligand-compatible use to immobilize with nearly any primary amine-containing compound or affinity ligand
Low non-specific binding the bead surface is pre-blocked and any nonreacted NHS-ester groups are fully quenched
Protocol-compatible protein coupling to the beads and downstream applications can be performed manually or by automation (e.g., Thermo Scientific KingFisher Instruments)
Recommended for:
Immobilization of ligands for the purification of recombinant proteins; Immobilization of antibodies for immunoprecipitations and co-immunoprecipitations free of antibody contamination in the eluates; Immobilization of secondary antibody for the affinity purification of antibody subtypes from ascites, serum and cell culture supernatant
Specifications
Upon receipt store at 4°C. Product shipped with an ice pack. | |
Liquid | |
1 mL | |
Amine |
NHS Ester | |
Pierce™ | |
Recombinant | |
Magnetic Affinity Bead |
Safety and Handling
- Pierce NHS-Activated Magnetic Beads
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- Acute toxicity Category 4
- Reproductive toxicity Category 1B
Hazard Statement
- H312-Harmful in contact with skin.
- H331-Toxic if inhaled.
- H360FD-May damage fertility. May damage the unborn child.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : N,N-dimethylacetamide
For Research Use Only. Not for use in diagnostic procedures.