missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ ONPG (o-nitrophenyl-β-D-galactopyranoside)
Preferred colorimetric beta-galactosidase (b-Gal) substrate for ELISA applications.
Brand: Thermo Scientific™ 34055
This item is not returnable.
View return policy
Description
Thermo Scientific™ Pierce ONPG Substrate is the preferred colorimetric substrate for ELISA applications involving β-galactosidase (β-Gal) as the reporter enzyme.
ONPG (ω-nitrophenyl-β-D-galactopyranoside) is the preferred colorimetric substrate for ELISA applications involving β-galactosidase (β-Gal) reporter enzyme. The enzyme-substrate reaction yields a yellow product with an absorbance maximum at 420nm but whose intensity is also very high at 405nm (matching a common plate reader filter set).Highlights:
- Product formed is completely soluble
- High extinction coefficient at 405nm
- Substrate yields a yellow product that is easily detectable in the visual range after stopping the reaction with 1M sodium carbonate

Specifications
ELISA Substrate | |
Colorimetric | |
Beta-Gal |
5 g | |
Chemical Substrate | |
Upon receipt store at -20°C protected from moisture and light. Product is shipped at ambient temperature. |
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
ZI390580 | Certificate of Analysis | 15/10/2024 | 34055 |
ZE390579 | Certificate of Analysis | 22/05/2024 | 34055 |
YL384650 | Certificate of Analysis | 25/01/2024 | 34055 |
YI381867 | Certificate of Analysis | 28/09/2023 | 34055 |
XK3619871A | Certificate of Analysis | 19/11/2022 | 34055 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ ONPG (o-nitrophenyl-β-D-galactopyranoside) > 5g
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction