missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peptide Desalting Spin Columns

Product Code. p-7113065 Shop All Thermo Scientific Products
Click to view available options
Quantity:
25 Columns
50 Columns
Unit Size:
Pack of 25
Pack of 50
This item is not returnable. View return policy

Product Code. 15808242

Brand: Thermo Scientific™ 89852

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

Pierce Peptide Desalting Spin Columns provide a convenient and reproducible way to desalt and remove contaminants from peptide samples prior to analysis by mass spectrometry.

Thermo Scientific Pierce Peptide Desalting Spin Columns are ready-to-use centrifuge spin columns that enable efficient desalting of peptide samples following enzymatic digestion. The polymer-based hydrophobic resin contained in each column provides excellent binding and recovery characteristics for peptide samples in preparation for mass spectrometry and other methods. Each spin column can bind from 5 μg to 5 mg of native and TMT-labeled peptides. The spin column format allows processing of multiple samples (10–300 μL each) in parallel in approximately 30 minutes.

Mass spectrometry is essential for studying biological compounds. However, many of the buffers and reagents used for sample preparation interfere with mass spectrometry analysis, leading to poor-quality spectra and reduced sensitivity. Pierce Peptide Desalting Spin Columns are designed to remove many of the interfering hydrophilic sample contaminants with minimal sample loss, thus enabling sensitive and comprehensive analysis of digested protein samples by mass spectrometry.

Remove MS-interfering contaminants—significantly reduces signal suppression and improves sensitivity in mass spectrometry
High binding capacity—hydrophobic polymer-based resin allows for recovery of peptide loads of native and TMT-labeled peptides from 5 μg to 5 mg
Easy to use—resin is provided in single-use spin column format that fits common 2-mL tubes
Compatible—resin is validated with a variety of complex samples, including TMT-labeled peptides, and is stable under extreme pH conditions

TRUSTED_SUSTAINABILITY

Specifications

Product Type Spin Desalting Column
Product Line Pierce
Content And Storage Store at 2–8°C.
Shipping Condition Room Temperature
Final Product Type Peptide
Detection Method Mass Spectrometry
Format Spin Column
Quantity 25 Columns
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.