missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Peroxide Assay Reagent B

Detect and measure hydrogen peroxide levels (H2O2) in biological samples using an iron (Fe) and xylenol orange (XO) reagent for microplates or tubes.

Brand:  Thermo Scientific™ 23282

Product Code. 11871355

  • £152.00 / 50mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C.
Peroxide Assay Reagent B
Pierce™
Not Target-Specific
Spectrophotometer, Microplate Reader
Pierce Peroxide Assay Reagent B
Colorimetric
Quantitative Peroxide Assay
Solution-based Detection, Absorbance
50 mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZG385409 Certificate TypeCertificate of Analysis Date27/08/2024 Catalog Number23282
Lot NumberZD391553 Certificate TypeCertificate of Analysis Date02/05/2024 Catalog Number23282
Lot NumberZA381925 Certificate TypeCertificate of Analysis Date28/02/2024 Catalog Number23282
Lot NumberYI381902 Certificate TypeCertificate of Analysis Date30/09/2023 Catalog Number23282
Lot NumberYG376135 Certificate TypeCertificate of Analysis Date23/08/2023 Catalog Number23282
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Peroxide Assay Reagent B > Reagent B; 50mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.