Learn More
Thermo Scientific™ Phenol:Chloroform + Tris Buffer
Perform DNA extractions with this phenol:chloroform:isoamyl alcohol mixture (25:24:1), accompanied by a buffer to equilibrate the solution at either pH 6.7 or pH 8.0.
£83.50 - £196.00
Specifications
Content And Storage | Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. |
---|---|
For Use With (Application) | DNA Extraction |
Product Type | Phenol Chloroform and Tris Buffer |
Form | Liquid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11886714
|
Thermo Scientific™
17908 |
100 mL |
£83.50
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11896714
|
Thermo Scientific™
17909 |
400 mL |
£196.00
400mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific™ Pierce Phenol:Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.
Thermo Scientific Pierce Phenol-Chloroform is phenol supplied premixed with chloroform:isoamyl alcohol (25:24:1) and accompanying bottle of buffer to equilibrate at either pH 6.7 or pH 8.0.
Highlights:
- Offers excellent, nuclease-free performance
- Phenol is supplied premixed with Chloroform:Isoamyl Alcohol (25:24:1)
- Supplied at pH 6.7 for special, lower pH DNA extractions; for most DNA applications, the alkaline buffer (included) should be added to produce a pH of 8.0
Specifications
Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. | |
DNA Extraction | |
Phenol Chloroform and Tris Buffer | |
Liquid |
Safety and Handling
- Phenol:Chloroform + Tris Buffer
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 2
- Acute toxicity Category 4
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Skin corrosion/irritation Category 1 B
- Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H331-Toxic if inhaled.
- H340-May cause genetic defects.
- H350-May cause cancer.
- H373-May cause damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Phenol, trichloromethane, 3-methylbutan-1-ol
For Research Use Only. Not for use in diagnostic procedures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.