missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Phenol, Saturated
Shop All Thermo Scientific Products
Click to view available options
Quantity:
400 mL
Unit Size:
400mL
Description
Thermo Scientific™ Pierce Phenol, Saturated is phenol supplied as a saturated solution (pH 6.6) and accompanied by a bottle of buffer to adjust the pH to 7.9 for typical molecular biology applications.
Phenol supplied as a saturated solution (pH 6.6) and accompanied by a bottle of buffer to adjust the pH to 7.9 for typical molecular biology applications.
Specifications
Specifications
| Content And Storage | Store in a segregated and approved area. Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, between the following temperatures: 2 to 8°C. |
| For Use With (Application) | Nucleic Acid Extraction |
| Product Type | Saturated Phenol |
| Quantity | 400 mL |
| Form | Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction