missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase
Periodate-aldehyde-activated horseradish peroxidase (HRP) enzyme and kit to label primary antibodies to obtain high-activity antibody-HRP conjugates.
£112.00 - £387.00
Specifications
Reactive Moiety | Aldehyde |
---|---|
Solubility | Water |
Chemical Reactivity | Amine |
Content And Storage | Note: EZ-Link Plus Activated Peroxidase contains 1-3 moles of aldehyde per mole of peroxidase. Storage: Upon receipt store product protected from moisture at -20°C. Product is shipped with an ice pack. |
Form | Solid |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10414224
|
Thermo Scientific™
31487 |
1mg |
£112.00
1mg |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
11849150
|
Thermo Scientific™
31488 |
5 x 1mg |
£387.00
Pack of 5 |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Thermo Scientific™ EZ-Link™ Plus Activated Peroxidase Kit contains horseradish peroxidase enzyme that has been aldehyde-activated with periodate to enable high-efficiency labeling and preparation of protein or antibody conjugates.
Other amine-reactive chemistries, like glutaraldehyde, have a tendency to polymerize and produce lower amounts of viable conjugate. Sugar residues present on HRP have been oxidized with periodate to produce aldehydes that react with primary amines. Plus Activated Peroxidase is mixed with the protein to be coupled at a pH compatible with your protein or antibody. After incubating the reaction for one to two hours, the conjugate linkage is reduced and the activated HRP quenched with ethanolamine. The conjugate is purified by desalting or by dialysis. If you are preparing an antibody-HRP conjugate, use the Thermo Scientific Pierce Conjugate Purification Kit (Part No. 44920) to separate unreacted enzyme from the conjugate.Highlights:
- Reacts with readily accessible primary amines to form a covalent secondary amine bond
- Can be stored for at least 12 months at -20°C
- Consistent conjugation yields reliable conjugates
- Enzyme activity is 120-200 units/mg
Specifications
Aldehyde | |
Amine | |
Solid | |
Enzyme Labeled |
Water | |
Note: EZ-Link Plus Activated Peroxidase contains 1-3 moles of aldehyde per mole of peroxidase. Storage: Upon receipt store product protected from moisture at -20°C. Product is shipped with an ice pack. |
|
EZ-Link™ | |
HRP (Horseradish Peroxidase) |
For Research Use Only. Not for use in diagnostic procedures.