missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Premium Grade EDC (1-ethyl-3-(3-dimethylaminopropyl)carbodiimide hydrochloride)

Product Code. 15245763 Shop All Thermo Scientific Products
Click to view available options
Quantity:
1 g
25 g
500 g
Unit Size:
1g
25g
500g
This item is not returnable. View return policy

Product Code. 15245763

Brand: Thermo Scientific™ PG82074

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Optimize applications where product integrity and risk minimization are paramount with this high-quality formulation of carbodiimide crosslinker.

Optimize applications where product integrity and risk minimization are paramount with the specially characterized Thermo ScientificTM PierceTM Premium Grade EDC, our highest quality formulation of this popular carbodiimide crosslinker.

  • The highest quality formulation of this popular carbodiimide crosslinker, specially characterized for applications where product integrity and risk minimization are paramount
  • Compared to the standard grade product, Premium Grade EDC provides more clearly defined quality and product support
  • A water-soluble carbodiimide crosslinker that activates carboxyl groups for spontaneous reaction with primary amines, enabling peptide immobilization and hapten-carrier protein conjugation
  • In the presence of N-hydroxysulfosuccinimide (sulfo-NHS), EDC can be used to convert carboxyl groups to amine-reactive sulfo-NHS esters
  • Store dessicated at -20°C

Features of Premium Grade EDC

  • High quality – identity and purity confirmed by several tests
  • Product integrity – enhanced level of testing and characterization compared to standard grade
  • Lot retention – ample supply of past lots retained to ensure future process testing
  • Change management – Change Control Notification (CCN) service
  • Consistent manufacture – batch-specific manufacturing documentation review

Applications

  • Conjugate carboxyl and amino groups among peptides and proteins
  • Couple haptens to immunogenic carrier proteins
  • Immobilize peptide antigens to affinity-purify antibodies
  • Create NHS-activated, amine-reactive labeling compounds
  • Crosslink proteins to carboxyl coated beads or surfaces
  • Activate nanoparticles with amine-reactive sulfo-NHS esters
TRUSTED_SUSTAINABILITY

Specifications

Chemical Reactivity Amine-Carboxyl
Cleavable No
Molecular Weight (g/mol) 191.7
PEGylated No
Spacer Arm Length 0.0 Å
Content And Storage Upon receipt store desiccated at -20°C.
Cell Permeability No
Shipping Condition Wet Ice
Product Line Pierce
Labeling Method Chemical Labeling
Crosslinker Type Heterobifunctional
Reactive Moiety Carbodiimide
Spacer Short (<10 Å)
Form Powder
Quantity 500 g
Solubility Water
Format Standard, Single-use, Premium-grade
Water Soluble Yes
Product Type Crosslinker
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.