missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A IgG Purification Kit, 1 mL

Purify antibodies or perform immunoprecipitation with this standard-capacity, immobilized native Protein A on beaded agarose resin.

Brand:  Thermo Scientific™ 44667

345.26 GBP valid until 2024-11-30
Use promo code "21651" to get your promotional price.


Product Code. 10056373

  • £566.00 / 1 set
Estimated Shipment: 09-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Perform a variety of antibody affinity purification methods with Thermo Scientific™ Pierce Protein A Agarose, a standard-capacity Protein A beaded agarose resin.

Pierce Protein A Agarose consists of purified native Protein A that has been covalently immobilized onto high-quality crosslinked 6% beaded agarose. The agarose beads have physical and chemical properties that enable them to be used in a variety of batch- or column-type affinity purification procedure. Several package formats are offered: 5mL and 25mL bottles of resin slurry, 1mL pre-packed columns and a complete IgG purification kit.

Protein A is a bacteria-derived protein that binds with high affinity and specificity to the Fc portion of antibodies, especially those of the IgG class. Compared to its alternative (Protein G), Protein A provides the highest binding capacity for subclasses of IgG from rabbits, pigs, dogs and cats. Protein A also has higher binding capacity for human IgG, except for IgG3, which it binds weakly. Protein A also binds weakly to mouse IgG1 and is not recommended for most applications with that antibody isotype.

Highlights:

  • Native Protein A – immobilized Protein A is ideal for polyclonal IgG purification from human, rabbit, pig, dog or cat serum

  • Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods

  • Inert and stable – superior manufacturing method immobilizes Protein A by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield

  • Standard capacity – Pierce Protein A Agarose has a normal load of immobilized Protein A, providing a binding capacity of 12 to 19mg human IgG/mL resin (approx. 6 to 8mg mouse IgG/mL resin)

Properties of crosslinked 6% beaded agarose (CL-6B):

  • Support pH Stability: 2 to 14 (short term); 3 to 13 (long term)
  • Average Particle Size: 45 to 165 microns
  • Exclusion Limit: 10,000 to 4,000,000 daltons
  • Maximum Volumetric Flow Rate: approx. 1mL/minute (for 1cm diameter column)
  • Maximum Linear Velocity: 30cm per hour
  • Maximum Pressure: less than 25psi (1.5 bar), defined as the maximum pressure drop across a column that the resin can withstand (Note: The indicated gauge pressure of a liquid chromatography apparatus may be measuring the total system pressure rather than the pressure drop across the column.)

Available Formats:

  • Resin Slurries: crosslinked 6% beaded agarose; 5mL and 25mL bottles
  • Spin Columns: 1mL columns
  • Purification Kit: complete kit containing five 1mL columns
  • Bulk and Custom: let us know your needs for bulk Pierce Protein A Agarose

Includes:

Kit contains Protein A Column (1mL each), Protein A IgG Binding Buffer, IgG Elution Buffer and desalting columns

TRUSTED_SUSTAINABILITY
Specifications

Specifications

IgG Purification Kit
Agarose Resin, Affinity
Protein A
6 to 8 mg Mouse IgG per Column
5 Column Kit
Pierce™
Kit, 15 mL Conical Spin Column
IgG Antibodies
Sufficient For: Purifying 6 to 8 mg mouse IgG per column
• Protein A Columns, 1 mL, 5 columns
• Protein A IgG Binding Buffer, 1 L
• IgG Elution Buffer, 500 mL
• Excellulose Desalting Columns, 5 mL, 5 columns

Store at 4°C.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.