missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein A/G Plus Agarose

Brand:  Thermo Scientific™ 20424

Additional Details : Weight : 0.01300kg

 View more versions of this product

Product Code. 15625315

  • £2965.00 / 20mL
Estimated Shipment: 26-07-2024
to see stock.
Add to basket

Description

Description

  • Protein A/G – immobilized recombinant fusion protein of the antibody-binding domains of Protein A and Protein G enables polyclonal IgG purification from nearly any mammalian species
  • Agarose resin – support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Inert and stable – superior manufacturing method immobilizes Protein A/G by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • High capacity – this 'Plus' variety of Pierce Protein A/G Agarose has a very dense load of immobilized Protein A/G, providing a binding capacity greater than 50 mg human IgG/mL resin
Specifications

Specifications

Agarose
IgG Antibodies, Ig Antibodies
Store in refrigerator (2–8°C).
Wet Ice
Protein A/G
Antibody Purification
Protein A/G, Agarose Resin
Agarose Bead
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.