missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Protein Concentrators PES, 30K MWCO

Thermo Scientific Pierce Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of proteins over 30 KDa, in sample volumes from 0.5 to 100 mL.

Brand:  Thermo Scientific™ 88531

233.22 GBP valid until 2025-05-31
Use promo code "25977" to get your promotional price.


 View more versions of this product

Product Code. 15451025

  • £249.00 / 24 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

Single-use
PES
5 to 20 mL
Protein
Room Temperature
Conical, Spin Column
30 kDa
Pierce™
24 Concentrators
Filter
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberZK400091 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88531
Lot NumberZK400091A Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88531
Lot NumberZK403222 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88531
Lot NumberZK403222A Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88531
Lot NumberZK408646 Certificate TypeCertificate of Analysis Date16/12/2024 Catalog Number88531
Special Offers

Special Offers

  • Multiple Manufacturers
    Advance your proteomics research with essential tools
    Promo Code: 25977

    Expires2025-05-31

    Days

    34

    Hours

    12

    Minutes

    23

    Seconds

    47
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Protein Concentrators PES, 30K MWCO > 5 to 20 mL, 24 Concentrators

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.