Learn More
Thermo Scientific™ Pierce™ Protein Concentrators PES, 50K MWCO, 0.5–100 mL
Thermo Scientific Pierce Protein Concentrators PES are easy-to-use centrifugal devices that provide fast processing and excellent recovery of proteins over 50 KDa, in sample volumes from 0.5 to 100 mL.
£78.00 - £306.00
Specifications
Filter Type Membrane Filter
Material (Membrane) PES
MWCO 50 kDa
Product Line Pierce™
Shipping Condition Room Temperature
View More Specs
Products 6
Product Code Brand Quantity Sample Volume Price Quantity & Availability  
Product Code Brand Quantity Sample Volume Price Quantity & Availability  
16320032
View Documents Promotion Details
Thermo Scientific™
88504
25 Concentrators ≤0.5 mL
£123.00
Pack of 25
16341964
View Documents Promotion Details
Thermo Scientific™
88538
10 Concentrators 2 to 6 mL
£78.00
Pack of 10
16351964
View Documents Promotion Details
Thermo Scientific™
88539
24 Concentrators 2 to 6 mL
£155.00
Pack of 24
16361964
View Documents Promotion Details
Thermo Scientific™
88540
10 Concentrators 5 to 20 mL
£156.00
Pack of 10
16371964
View Documents Promotion Details
Thermo Scientific™
88541
24 Concentrators 5 to 20 mL
£306.00
Pack of 24
16381964
View Documents Promotion Details
Thermo Scientific™
88542
4 Concentrators 20 to 100 mL
£178.00
Pack of 4
Description
Show More
Specifications
Specifications
Membrane Filter
50 kDa
Room Temperature
PES
Pierce™
Protein Concentrator
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Pierce™ Protein Concentrators PES, 50K MWCO, 0.5–100 mL