missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Protein-Free Blocking Buffer
Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background.
£58.00 - £197.00
Specifications
Product Type | Blocking Buffer, protein-free |
---|---|
Form | Liquid |
Concentration | 1X |
Blocking Agent | Non-protein compound |
Content And Storage | Upon receipt store product at room temperature. After opening, store product at 4°C. |
Product Code | Brand | Description | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity & Availability | |||||
13474299
|
Thermo Scientific™
37584 |
Protein-Free (PBS) Blocking Buffer, 100 mL |
£58.50
100mL |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10229663
|
Thermo Scientific™
37572 |
Protein-Free (PBS) Blocking Buffer, 1 L |
£193.00
1 L |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13484299
|
Thermo Scientific™
37585 |
Protein-Free (TBS) Blocking Buffer, 100 mL |
£58.00
100mL |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10269613
|
Thermo Scientific™
37570 |
Protein-Free (TBS) Blocking Buffer, 1 L |
£185.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10147483
|
Thermo Scientific™
37573 |
Protein-Free T20 (PBS) Blocking Buffer, 1 L |
£194.00
1 L |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10137483
|
Thermo Scientific™
37571 |
Protein-Free T20 (TBS) Blocking Buffer, 1 L |
£197.00
1 L |
Estimated Shipment: 25-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Pierce Protein-Free Blocking Buffer is a non-protein compound that provides effective blocking for membrane-based and plate-based protein detection methods, resulting in extremely low background. It is supplied in a convenient, ready-to-use format.
Features of Protein-Free Blocking Buffer include:• Protein-free blocker—minimizes or eliminates cross-reactivity associated with protein-based blocking buffers
• Applications—compatible with western blotting (membranes), ELISA (microplates), and arrays (coated glass slides)
• Streptavidin-friendly—free of biotin; no interference with avidin-biotin detection systems
• High-performance—optimized and validated in many protein methods to provide high signal-to-noise ratio (i.e., no quenching of specific binding and signal but eliminating nonspecific binding and background)
Traditional blocking buffers contain proteins that can cross-react in certain immunodetection systems, resulting in high background and reduced signal. Pierce Protein-Free Blocking Buffers are devoid of protein while remaining highly effective at blocking plates, membranes, and other surfaces for ELISA, western blotting, glass slide arrays, and other applications.
Specifications
Blocking Buffer, protein-free | |
1X | |
Upon receipt store product at room temperature. After opening, store product at 4°C. |
Liquid | |
Non-protein compound |