missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Rapid Gel Clot Endotoxin Assay Kits

Rapid Gel Clot Endotoxin Assay Kits provide rapid and easy-to-read results for presence or absence of gram-negative bacterial endotoxin in the test sample. The kit delivers a qualitative 'Pass' or 'Fail' result, with kit sensitivity options ranging from 0.03 to 0.5 EU/mL, each supporting 10 tests.

Brand:  Thermo Scientific™ A43882

77.33 GBP valid until 2025-03-29
Use promo code "24111" to get your promotional price.


 View more versions of this product

Product Code. 17841648

  • £116.00 / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers

Alert:

To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price

This item is not returnable. View return policy

Description

Description

Endotoxin is a lipopolysaccharide (LPS) that constitutes much of the outer membrane of the cell wall in gram-negative bacteria. LPS is the most abundant antigen on the cell surface of most gram-negative bacteria. Thermo Fisher Scientific Pierce Rapid Gel Clot Endotoxin Assay kits provide rapid and easy-to-read qualitative results indicating the presence or absence of endotoxins. They are best suited for samples that are viscous or highly colored and are available in a variety of sensitivities: 0.03, 0.125, 0.25, and 0.5 EU/mL.

These Rapid Gel Clot Endotoxin Assay kits have a fast, efficient protocol, are reproducible and contain all the reagents and supplies such as positive control tubes, endotoxin-free tubes, and pipette tips required for each test. Each kit is sufficient for 10 tests, using 0.5 mL of test sample.

Pierce Rapid Gel Clot Endotoxin Assay kits use amebocyte lysates derived from the Limulus horseshoe crab hemolymph to detect endotoxin levels in samples. Limulus amebocyte lysate (LAL) is widely used as a simple and sensitive assay for detection of endotoxins. When endotoxin encounters the amebocyte lysate, a series of enzymatic reactions form a gel-like clot. In a positive test, a clot will form in the sample tube, indicating that the amount of endotoxin in the sample is greater than or equal to the listed sensitivity (in EU/mL) of the kit. A lack of gel clot formation in the tube is considered a negative result, indicative of an endotoxin concentration in the test tube below the kit’s sensitivity.

Endotoxins are released in their surrounding environment when gram-negative bacteria undergo normal cell division or in large concentrations when they are damaged or undergo cell lysis. Given endotoxins are pyrogenic (induce fever) in mammals, identifying and monitoring endotoxin levels is extremely important in many fields. Endotoxin testing of raw materials, intermediates, and final drug products is essential for patient safety. Each kit lot has a defined assay incubation time that is listed in the Certificate of Analysis accompanying each kit. This kit is not licensed by the FDA and is not to be used for the end-product release of pharmaceutical drugs, devices, or biologics that are FDA regulated.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

• Positive Control Tubes
• Endotoxin-Free Sample Reaction Tubes
• Endotoxin-Free Pipette Tips
• Endotoxin-Free Sample Bottles

Store Sample and Positive Control Reaction Tubes at 4°C.
Store pipette tips and sample bottles at room temperature.

Kit
This kit is not licensed by the FDA and is not to be used for the end-product release of pharmaceutical drugs, devices, or biologics that are FDA regulated. For Research Use Only.
10 Assays
Pierce™ Rapid Gel Clot Endotoxin Assay Kit, 0.50 EU/mL
End-Point Gel Clot LAL Assay
Pierce™
0.5 EU/mL
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

4 results found
Lot Number Certificate Type Date Catalog Number
Lot Number0BC8T00900 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberA43882
Lot Number0C86Q80500 Certificate TypeCertificate of Analysis Date29/09/2023 Catalog NumberA43882
Lot Number0C23T50800 Certificate TypeCertificate of Analysis Date29/07/2023 Catalog NumberA43882
Lot NumberSPECS Certificate TypeCertificate of Analysis Date10/06/2023 Catalog NumberA43882
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Pierce™ Rapid Gel Clot Endotoxin Assay Kits > 0.50 EU/mL, 10 tests

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.