Learn More
Thermo Scientific™ ELISA Reagent Reservoir
Rapidly dispense reagents with multi-channel pipettors using these sterile, sturdy and disposable reservoirs molded from high-impact polystyrene.
Brand: Thermo Scientific™ 15075
207.10 GBP valid until 2024-08-31
Use promo code "21743" to get your promotional price.
Code : PN
Additional Details : Weight : 1.81440kg
Description
Thermo Scientific™ Pierce Reagent Reservoirs are sterile, sturdy, disposable and molded from high-impact polystyrene.
Additional Information and Online Ordering »
Highlights:
- Sterile, sturdy reservoirs are molded from high-impact polystyrene
Reservoirs facilitate repeated pick up of reagents by multi-channel pipettors for delivery to 96-well plates
Specifications
Pierce ELISA Reagent Reservoirs | |
Polystyrene | |
Reagent Reservoir | |
Store at room temperature. | |
Single-use |
50 mL | |
Sterile | |
200 Units | |
96-Well Plate, 384-Well Plate | |
V-bottom |
For Research Use Only. Not for use in diagnostic procedures.