missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Recombinant Protein G
Detect mouse and human antibodies, especially IgG isotypes, in immunoassays with this purified, biotinylated or peroxidase-conjugated recombinant Protein G.
£446.00 - £2245.00
Specifications
Product Line | Pierce™ |
---|---|
Form | Powder |
Protein Subtype | Protein G |
Target | Antibodies |
Content And Storage | Upon receipt store at -20°C. Product is shipped at ambient temperature. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10712725
|
Thermo Scientific™
21193 |
5 mg |
£446.00
5mg |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15285683
|
Thermo Scientific™
77675 |
50 mg |
£904.00
50mg |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
15295683
|
Thermo Scientific™
77676 |
500 mg |
£2245.00
500mg |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein G enable probing and detection of mouse and human antibodies, especially IgG isotypes, in Western blotting, ELISA, IHC and other immunoassay protocols.
Protein G is a bacterial cell wall protein isolated from group G Streptococci. DNA sequencing of native Protein G identifies two IgG-binding domains and sites for albumin and cell surface binding. The albumin and cell surface binding domains have been eliminated from Recombinant Protein G to reduce nonspecific binding and, therefore, can be used to separate IgG from crude samples. Optimal binding occurs at pH 5, although binding is also effective at pH 7.0 to 7.2.Highlights:
- Contains two Fc-binding domains per protein
- Better than Protein A for mouse (incl. IgG1), human (incl. IgG3) and rat, goat and cow antibodies
- Poorer than Protein A for guinea pig, pig, dog and cat
- Does not bind human IgM, IgD or IgA
Specifications
Pierce™ | |
Protein G | |
Upon receipt store at -20°C. Product is shipped at ambient temperature. |
Powder | |
Antibodies |
For Research Use Only. Not for use in diagnostic procedures.