missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Recombinant Protein L, Peroxidase Conjugated

Product Code. 10321504 Shop All Thermo Scientific Products
Click to view available options
Quantity:
0.5 mg
Unit Size:
0.5mg
This item is not returnable. View return policy

Product Code. 10321504

Brand: Thermo Scientific™ 32420

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Detect mouse and human antibodies containing kappa light chains in immunoassays using this purified, biotinylated or peroxidase-conjugated, recombinant Protein L.

Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein L enable probing and detection of mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.

Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.

Highlights:

  • Contains four immunoglobulin-binding domains per protein
  • Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
  • Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains
TRUSTED_SUSTAINABILITY

Specifications

Product Type Protein L, Peroxidase Conjugate
Conjugate HRP
Quantity 0.5 mg
Product Line Pierce
Form Powder
Protein Subtype Protein L
Target Antibodies
Content And Storage Store in original container protected from direct sunlight in a dry, cool and well-ventilated area, but do not store above -20°C.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.