missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Pierce™ Recombinant Protein L
Detect mouse and human antibodies containing kappa light chains in immunoassays using this purified, biotinylated or peroxidase-conjugated, recombinant Protein L.
£183.00 - £7420.00
Specifications
Product Line | Pierce™ |
---|---|
Form | Powder |
Protein Subtype | Protein L |
Target | Antibodies |
Content And Storage | Upon receipt store at 4°C. Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C. |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10658464
|
Thermo Scientific™
21189 |
1 mg |
£183.00
1mg |
Estimated Shipment: 30-07-2024
Log in to see stock. |
|||||
15225693
|
Thermo Scientific™
77679 |
50 mg |
£1832.00
50mg |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
15235693
|
Thermo Scientific™
77680 |
500 mg |
£7420.00
500mg |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
Purified and conjugated forms of Thermo Scientific™ Pierce Recombinant Protein L enable probing and detection of mouse and human antibodies containing kappa light chains in Western blotting, ELISA, IHC and other immunoassay protocols.
Protein L is an immunoglobulin-binding protein that was originally derived from the bacteria Peptostreptococcus magnus, but is now produced recombinantly. Protein L has the unique ability to bind through kappa light chain interactions without interfering with an antibody's antigen-binding site. This gives Protein L the ability to bind a wider range of Ig classes and subclasses than other antibody-binding proteins. Protein L will bind to all classes of Ig (IgG, IgM, IgA, IgE and IgD). Protein L will also bind single chain variable fragments (scFv) and Fab fragments. Protein L binds kappa I, III, and IV in human and kappa I on mouse.Highlights:
- Contains four immunoglobulin-binding domains per protein
- Binds members of all classes of Ig (IgG, IgM, IgA, IgE and IgD) that contain kappa light chains
- Binds single chain variable fragments (scFv) and Fab fragments that contain kappa light chains
Specifications
Pierce™ | |
Protein L | |
Upon receipt store at 4°C. Note: Store reconstituted product for up to one month at 4°C. For long-term storage, aliquot, add an equal volume of glycerol and store at -20°C. |
Powder | |
Antibodies |
For Research Use Only. Not for use in diagnostic procedures.