Learn More
Thermo Scientific™ Pierce™ Renilla Luciferase Flash Assay Kit
Assay regulatory element transcriptional activity in whole cell lysates with this highly sensitive flash-based Renilla luciferase kit.
£115.00 - £584.00
Specifications
Type | Assay Kit |
---|---|
Target | Luciferase, Green Renilla Luciferase |
Assay | Reporter Enzyme, Luciferase Reporter Assay |
Compatible Cells | Mammalian Cells |
Detection Method | Bioluminescent |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12162240
|
Thermo Scientific™
16164 |
100 Reaction Kit |
£115.00
Each |
Estimated Shipment: 30-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12137850
|
Thermo Scientific™
16165 |
1000 Reaction Kit |
£584.00
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
The Thermo Scientific™ Pierce Renilla Luciferase Flash Assay Kit provides researchers with a highly sensitive intracellular assay for transcriptional activity of regulatory elements in mammalian whole cell lysate.
This Flash Assay Kit contains reagents for measuring the activity of Renilla luciferase in mammalian cell lysates. When used with Thermo Scientific Green Renilla Luc Plasmids, the kit provides an extremely sensitive bioluminescent reporter assay system for intracellular detection of promoter or pathway activity. The signal produced by Green Renilla Luc is considerably greater than signal from either native firefly or Renilla luciferases assayed under similar conditions. Light output captured using a luminometer can be correlated with the amount of Renilla luciferase protein produced and used to determine the activity of the promoter driving Renilla expression.Highlights:
Sensitive – greater sensitivity allows utilization of smaller numbers of cells
Cost effective – highest sensitivity assays result in decreased reagent consumption
Time-saving – assays require minimal sample handling
Automation-friendly – amenable to high throughput screens
Convenient – contains a universal cell lysis buffer and optimized flash assay reagent
Safe – allows one to perform non-radioactive assays
Includes:
Cell lysis buffer, reaction buffer and substrate
Requires:
Renilla luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan Ascent and Varioskan Flash Microplate Readers
Recommended for:
Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies
Specifications
Assay Kit | |
Reporter Enzyme, Luciferase Reporter Assay | |
Bioluminescent | |
384-well plate, 96-well plate | |
Chemical Substrate | |
Luciferase Substrate | |
Coelenterazine |
Luciferase, Green Renilla Luciferase | |
Mammalian Cells | |
Luminometer (Microplate) | |
Enzyme Labeled | |
Enhanced Chemiluminescence | |
Pierce™ |
For Research Use Only. Not for use in diagnostic procedures.