missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Pierce™ Renilla Luciferase Glow Assay Kit

Product Code. p-4532169 Shop All Thermo Scientific Products
Click to view available options
Quantity:
100 Reaction Kit
1000 Reaction Kit
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12172240

Brand: Thermo Scientific™ 16166

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Obtain bright and stable bioluminescence signal (half-life approx. 3 hour) in the presence of Renilla luciferase reporters.

Thermo Scientific™ Pierce Renilla Luciferase Glow Assay Kit provides an extremely bright and stable bioluminescence signal (half-life approx. 3 hours), especially in the presence of Green Renilla luciferase reporter.

This Pierce Luciferase Glow Assay Kit contains reagents for measuring the activity of Renilla luciferase in cell lysates. In the presence of luciferase, the glow reagent provides stable bioluminescent signal and is highly suited for luminometers without injectors or for batch processing of samples. The light output generated by the luciferase reaction can be correlated with the amount of luciferase protein produced which in turn is proportional to the promoter activity driving the luciferase expression. The glow assay reagents were optimized for best results with Thermo Scientific Green Renilla Luc Plasmids; however, this glow kit may be used to detect the activity of other Renilla luciferase derivatives that use coelenterazine as a substrate.

Highlights:

  • Sensitive – highly sensitive detection of Green Renilla luciferase activity

  • Stable – increased signal stability compared to flash assays

  • Simple – does not require a luminometer with injectors

  • Compatible – assay reagents compatible with other Renilla luciferases

  • Automation-friendly – amenable to high throughput assays

  • Convenient – contains a universal cell lysis buffer and optimized glow assay reagent

  • Safe – allows one to perform non-radioactive assays

Includes:

Cell lysis buffer, reaction buffer and substrate

Requires:

Renilla luciferase and luminometer or other instrument capable of monitoring luminescence, such as Thermo Scientific Luminoskan™ Ascent and Varioskan™ Flash Microplate Readers

Recommended for:

Promoter studies for analyzing cis- regulatory elements and trans-acting factors; Drug screening; siRNA and miRNA screening; Multiplexed assays to study off-target effects; Protein localization reporter assays; Signal transduction pathway analysis; RNA splicing studies

TRUSTED_SUSTAINABILITY

Specifications

Type Assay Kit
Target Luciferase, Green Renilla Luciferase
Quantity 100 Reaction Kit
Content And Storage Upon receipt store kit at -80°C or store individual components as indicated above.
• Renilla Glow Assay Buffer, 5 mL, store at 4°C
• Coelenterazine (100X), 50 μL, store at -80°C
• 2X Cell Lysis Buffer, 6 mL, store at room temperature
Assay Reporter Enzyme, Luciferase Reporter Assay
Compatible Cells Mammalian Cells
Detection Method Bioluminescence
For Use With (Equipment) Luminometer (Microplate)
Format 384-well plate, 96-well plate
Label Type Enzyme Labeled
Substrate Properties Chemical Substrate
Technique Enhanced Chemiluminescence
Substrate Type Luciferase Substrate
Product Line Pierce
Substrate Coelenterazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Pierce Renilla Luciferase Glow Assay Kit
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Flammable liquid Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : methanol

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.