Learn More
Thermo Scientific™ Pierce™ Reserpine Standard for LC-MS
An exact solution of reserpine specifically designed for performance evaluation of mass spectrometers, including the Thermo Scientific™ ion trap and TSQ series of instruments.
Brand: Thermo Scientific™ 88326
Code : PN
Additional Details : CAS Number : 67-63-0 Weight : 2.26800kg Transport : UN number : 1993 Chem class : 3 Pack group : II
Description
Thermo Scientific™ Pierce Reserpine Standard for LC-MS is an exact solution of reserpine specifically designed for performance evaluation of mass spectrometers, including the Thermo Scientific ion trap and TSQ series of instruments.
Additional Information and Online Ordering »
The Pierce Reserpine Standard for LC-MS is a pre-diluted liquid formulation ideal to quickly perform installation specifications of Thermo Scientific (and other) mass spectrometers. The standard is provided in a concentration of 100pg/μL in 50% isopropyl alcohol and requires minimal additional dilutions. The product is provided in a pack of 5 x 1mL glass amber vials with PTFE-lined screw caps. The Pierce Reserpine Standard for LC-MS is manufactured at an ISO 9001 facility and each lot is quality controlled with strict specifications.
Highlights:
Convenient – provided pre-diluted and requires minimal additional dilutions to reach target concentration for injection
Safe handling – unlike glass ampules, cap can be easily removed and replaced when withdrawing solution
High-purity – mass spectrometry grade reagent in non-leachable screw-cap vials
Stable – store at 4°C for up to one year
Specifications
Mass Spectrometer | |
Pierce™ | |
Peptides, Small Molecules | |
Mass Spectrometry | |
Liquid |
Reserpine Standard | |
Store in a cool, dry, well-ventilated area, protected from direct sunlight. | |
Peptides, Small Molecules | |
Calibration |
Safety and Handling
- Pierce Reserpine Standard for LC-MS
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : propan-2-ol
For Research Use Only. Not for use in diagnostic procedures.